Learn More
STEMCELL Technologies EasySep™ HLA B Cell Enrichment: Complete Processing Kit for Whole Blood, For processing: 200 mL whole blood processed
SDP
Catalog No. 501974734
Encompass
The EasySep™ HLA B Cell Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human B cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD33, CD36, CD41, CD43, CD56, CD66b, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. 50-197-4734 Supplier STEMCELL Technologies Supplier No. 19954HLA
The EasySep™ HLA B Cell Enrichment Kit: Complete Processing Kit for Whole Blood is designed to isolate human B cells from HetaSep™-treated whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD33, CD36, CD41, CD43, CD56, CD66b, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.