Learn More
STEMCELL Technologies EasySep™ HLA B Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974671
Encompass
The EasySep™ HLA B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet and without the use of columns. Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays.
Catalog No. 50-197-4671 Supplier STEMCELL Technologies Supplier No. 19054HLA
The EasySep™ HLA B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet and without the use of columns. Desired cells are poured off into a new tube and are ready for serology or flow cytometry crossmatch assays.