Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit, For processing: 60 mL buffy coat or whole blood processed
SDP
Catalog No. 501974572
Encompass
The EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit is designed to isolate CD3+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on T cells and NKT cells.This product replaces the EasySep™ Human Whole Blood CD3 Positive Selection Kit (Catalog #18081) and EasySep™ HLA Whole Blood CD3 Positive Selection Kit (Catalog #18081HLA), for even faster cell isolations.
Catalog No. 50-197-4572 Supplier STEMCELL Technologies Supplier No. 17871
The EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit is designed to isolate CD3+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on T cells and NKT cells.This product replaces the EasySep™ Human Whole Blood CD3 Positive Selection Kit (Catalog #18081) and EasySep™ HLA Whole Blood CD3 Positive Selection Kit (Catalog #18081HLA), for even faster cell isolations.