Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit, For processing: 60 mL buffy coat or whole blood processed
SDP
Catalog No. 501974586
Encompass
The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD15 antigen is expressed on granulocytes and monocytes.This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit (Catalog #18681) and EasySep™ HLA Whole Blood CD15 Positive Selection Kit (Catalog #18681HLA), for even faster cell isolations.
Catalog No. 50-197-4586 Supplier STEMCELL Technologies Supplier No. 17881
The EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit is designed to isolate highly purified CD15+ cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD15 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD15 antigen is expressed on granulocytes and monocytes.This product replaces the EasySep™ Human Whole Blood CD15 Positive Selection Kit (Catalog #18681) and EasySep™ HLA Whole Blood CD15 Positive Selection Kit (Catalog #18681HLA), for even faster cell isolations.