Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit, For processing: 60 mL whole blood processed
SDP
Supplier:  STEMCELL Technologies 17885
Encompass
The EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit is designed to isolate highly purified myeloid (including CD14+ and CD66b+) cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 antigen is expressed on myeloid progenitor cells, granulocytes and monocytes.This product replaces the EasySep™ Human Whole Blood CD33 Positive Selection Kit (Catalog #18287) and EasySep™ HLA Whole Blood CD33 Positive Selection Kit (Catalog #18287HLA) for even faster cell isolations.
Catalog No. 50-197-4592