Learn More
STEMCELL Technologies EasySep HLA Chimerism Whole Bl, 60 mL buffy coat or whole blood processed
SDP
Catalog No. 502572382
Encompass
Column-free immunomagnetic positive selection of human B cells for HLA analysis
Catalog No. 50-257-2382 Supplier STEMCELL Technologies Supplier No. 17886
Column-free immunomagnetic positive selection of human B cells for HLA analysis