Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood Myeloid Positive Selection Kit, For processing: 60 mL buffy coat or whole blood processed
SDP
Catalog No. NC1456781
Encompass
Isolate highly purified CD33+ and CD66b+ myeloid cells, including myeloid progenitor cells, granulocytes, and monocytes, from fresh whole blood prior to chimerism testing. Desired cells are targeted with tetrameric antibody complexes recognizing CD33, CD66b, and dextran-coated magnetic particles, and separated without the use of columns through immunomagnetic positive selection using an EasySep™ magnet. Cells of interest remain in the tube while unwanted cells are poured off. This product can be used with RoboSep™ to automate immunomagnetic cell isolation. Visit www.EasySep.com for more information.
Catalog No. NC1456781 Supplier STEMCELL Technologies Supplier No. 17884
Isolate highly purified CD33+ and CD66b+ myeloid cells, including myeloid progenitor cells, granulocytes, and monocytes, from fresh whole blood prior to chimerism testing. Desired cells are targeted with tetrameric antibody complexes recognizing CD33, CD66b, and dextran-coated magnetic particles, and separated without the use of columns through immunomagnetic positive selection using an EasySep™ magnet. Cells of interest remain in the tube while unwanted cells are poured off. This product can be used with RoboSep™ to automate immunomagnetic cell isolation. Visit www.EasySep.com for more information.