Learn More
STEMCELL Technologies EasySep™ Human Resting CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1254490
Encompass
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Catalog No. NC1254490 Supplier STEMCELL Technologies Supplier No. 17962
The EasySep™ Human Resting CD4+ T Cell Isolation Kit is designed to isolate resting CD4+ T cells from fresh or previously frozen human peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.