Learn More
STEMCELL Technologies EasySep™ Human Basophil Isolation Kit, For isolating: 1 x 10^9 cells
SDP
Catalog No. NC1611146
Encompass
The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood.This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations.
Catalog No. NC1611146 Supplier STEMCELL Technologies Supplier No. 17969
The EasySep™ Human Basophil Isolation Kit is designed to isolate basophils from a polymorphonuclear cell-rich fraction by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-basophils and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation of fresh peripheral blood.This product replaces the EasySep™ Human Basophil Enrichment Kit (Catalog #19069) for even faster cell isolations.