Learn More
STEMCELL Technologies EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0187712
Encompass
The EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion is designed to isolate monocytes and CD14+CD16+ monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The CD14+CD16+ subset of monocytes (~10% in blood of healthy individuals) has characteristics of tissue macrophages and expands greatly in acute and chronic inflammatory disease. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-monocyte cells and magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For applications in which removal of all CD16+ cells is desired, we recommend the EasySep™ Human Monocyte Enrichment Kit (/products/easysep-human-monocyte-isolation-kit.html) (Catalog #19359), which contains anti-CD16.
Catalog No. NC0187712 Supplier STEMCELL Technologies Supplier No. 19058
The EasySep™ Human Monocyte Enrichment Kit without CD16 Depletion is designed to isolate monocytes and CD14+CD16+ monocytes from fresh or previously frozen peripheral blood mononuclear cells by negative selection. The CD14+CD16+ subset of monocytes (~10% in blood of healthy individuals) has characteristics of tissue macrophages and expands greatly in acute and chronic inflammatory disease. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-monocyte cells and magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For applications in which removal of all CD16+ cells is desired, we recommend the EasySep™ Human Monocyte Enrichment Kit (/products/easysep-human-monocyte-isolation-kit.html) (Catalog #19359), which contains anti-CD16.