Learn More
STEMCELL Technologies EasySep™ Human Naïve CD8+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1280859
Encompass
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD36, CD45RO, CD56, CD57, CD94, CD123, CD244, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the EasySep™ Human Naïve CD8+ T Cell Isolation Kit II (17968) (https://www.stemcell.com/products/easysep-human-naive-cd8-t-cell-isolation-kit-ii.html) which isolates cells in as little as 11 minutes.
This item is not returnable. View return policy
Catalog No. NC1280859 Supplier STEMCELL Technologies Supplier No. 19258
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD4, CD14, CD16, CD19, CD20, CD36, CD45RO, CD56, CD57, CD94, CD123, CD244, TCRγ/δ, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the EasySep™ Human Naïve CD8+ T Cell Isolation Kit II (17968) (https://www.stemcell.com/products/easysep-human-naive-cd8-t-cell-isolation-kit-ii.html) which isolates cells in as little as 11 minutes.