Learn More
STEMCELL Technologies EasySep™ Human CD2 Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974589
Encompass
The EasySep™ Human CD2 Positive Selection Kit II is designed to isolate CD2+ cells from fresh or previously frozen human peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD2 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD2 antigen is expressed on T cells and NK cells.This product replaces the EasySep™ Human CD2 Positive Selection Kit (Catalog #18657) for even faster cell isolations.
Catalog No. 50-197-4589 Supplier STEMCELL Technologies Supplier No. 17883
Add to Cart
Add to Cart
The EasySep™ Human CD2 Positive Selection Kit II is designed to isolate CD2+ cells from fresh or previously frozen human peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD2 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD2 antigen is expressed on T cells and NK cells.This product replaces the EasySep™ Human CD2 Positive Selection Kit (Catalog #18657) for even faster cell isolations.