Learn More
STEMCELL Technologies EasySep™ Human CD3 Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0883710
Encompass
The EasySep™ Human CD3 Positive Selection Kit II is designed to isolate CD3+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on all T cells and CD56+ NKT cells.This product replaces the EasySep™ Human CD3 Positive Selection Kit (Catalog #18051) for even faster cell isolations.
Catalog No. NC0883710 Supplier STEMCELL Technologies Supplier No. 17851
The EasySep™ Human CD3 Positive Selection Kit II is designed to isolate CD3+ cells from fresh or previously frozen peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD3 antigen is expressed on all T cells and CD56+ NKT cells.This product replaces the EasySep™ Human CD3 Positive Selection Kit (Catalog #18051) for even faster cell isolations.