Learn More
STEMCELL Technologies EasySep™ Human CD33 Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1604385
Encompass
The EasySep™ Human CD33 Positive Selection Kit II is designed to isolate CD33+ cells from ammonium chloride lysed blood or bone marrow, or from nucleated cells prepared by density separation with RosetteSep™ DM-M density medium (Catalog #15725) by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 antigen is expressed on cells of the myeloid lineage.This product replaces the EasySep™ Human CD33 Positive Selection Kit (Catalog #18257) for even faster cell isolations.
Catalog No. NC1604385 Supplier STEMCELL Technologies Supplier No. 17876
The EasySep™ Human CD33 Positive Selection Kit II is designed to isolate CD33+ cells from ammonium chloride lysed blood or bone marrow, or from nucleated cells prepared by density separation with RosetteSep™ DM-M density medium (Catalog #15725) by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 antigen is expressed on cells of the myeloid lineage.This product replaces the EasySep™ Human CD33 Positive Selection Kit (Catalog #18257) for even faster cell isolations.