Learn More
STEMCELL Technologies EasySep™ Human Gamma/Delta T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0782149
Encompass
The EasySep™ Human Gamma/Delta T Cell Isolation Kit is designed to isolate gamma/delta T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-gamma/delta T cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube. To obtain a custom kit optimized for culture-expanded PBMCs, contact STEMCELL Technologies’ Technical Support at techsupport@stemcell.com.
Catalog No. NC0782149 Supplier STEMCELL Technologies Supplier No. 19255
The EasySep™ Human Gamma/Delta T Cell Isolation Kit is designed to isolate gamma/delta T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-gamma/delta T cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube. To obtain a custom kit optimized for culture-expanded PBMCs, contact STEMCELL Technologies’ Technical Support at techsupport@stemcell.com.