Learn More
STEMCELL Technologies EasySep™ Human ILC2 Isolation Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. 501974563
Encompass
The EasySep™ Human ILC2 Isolation Kit is designed to isolate Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic positive selection without the use of a flow sorter.First, CRTH2+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the EasySep™-isolated CRTH2+ cells, and unwanted non-ILC2s are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction contains highly purified ILC2s that are immediately ready for downstream applications.Following cell isolation with this EasySep™ kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Catalog No. 50-197-4563 Supplier STEMCELL Technologies Supplier No. 17782
The EasySep™ Human ILC2 Isolation Kit is designed to isolate Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic positive selection without the use of a flow sorter.First, CRTH2+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the EasySep™-isolated CRTH2+ cells, and unwanted non-ILC2s are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction contains highly purified ILC2s that are immediately ready for downstream applications.Following cell isolation with this EasySep™ kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.