Learn More
STEMCELL Technologies EasySep™ Human Myeloid Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1647595
Encompass
The EasySep™ Human Myeloid Positive Selection Kit II is designed to isolate CD33+ and CD66b+ cells from ammonium chloride lysed blood or bone marrow by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33, CD66b and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 and CD66b antigens are expressed on myeloid progenitors monocytes and granulocytes.This product replaces the EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) for even faster cell isolations.
Catalog No. NC1647595 Supplier STEMCELL Technologies Supplier No. 17893
The EasySep™ Human Myeloid Positive Selection Kit II is designed to isolate CD33+ and CD66b+ cells from ammonium chloride lysed blood or bone marrow by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33, CD66b and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 and CD66b antigens are expressed on myeloid progenitors monocytes and granulocytes.This product replaces the EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) for even faster cell isolations.