Learn More
STEMCELL Technologies EasySep™ Human Naïve CD8+ T Cell Isolation Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1605234
Encompass
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit II is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically-labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This product replaces the EasySep™ Human Naïve CD8+ T Cell Enrichment Kit (Catalog #19158) for even faster cell isolations.
This item is not returnable. View return policy
Catalog No. NC1605234 Supplier STEMCELL Technologies Supplier No. 17968
The EasySep™ Human Naïve CD8+ T Cell Isolation Kit II is designed to isolate naïve CD8+ T cells from fresh peripheral blood mononuclear cells by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically-labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This product replaces the EasySep™ Human Naïve CD8+ T Cell Enrichment Kit (Catalog #19158) for even faster cell isolations.