Learn More
STEMCELL Technologies EasySep™ Human Neutrophil Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974615
Encompass
The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation or red blood cell lysis.This product replaces the EasySep™ Human Neutrophil Enrichment Kit (Catalog #19257), for even faster cell isolations.
Catalog No. 50-197-4615 Supplier STEMCELL Technologies Supplier No. 17957
The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation or red blood cell lysis.This product replaces the EasySep™ Human Neutrophil Enrichment Kit (Catalog #19257), for even faster cell isolations.