Learn More
STEMCELL Technologies EasySep™ Human Pan-B Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1142093
Encompass
The EasySep™ Human Pan-B Cell Enrichment Kit is designed to isolate B cells including plasma cells from normal fresh or previously frozen peripheral blood mononuclear cells by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD36, CD42b, CD56, CD66b, CD123, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. NC1142093 Supplier STEMCELL Technologies Supplier No. 19554
The EasySep™ Human Pan-B Cell Enrichment Kit is designed to isolate B cells including plasma cells from normal fresh or previously frozen peripheral blood mononuclear cells by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD36, CD42b, CD56, CD66b, CD123, glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.