Learn More
STEMCELL Technologies EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit, For processing: 20 mL biofluid processed
SDP
Catalog No. 501974600
Encompass
The EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection. Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD9, CD81, and CD63, and magnetic particles. Labeled extracellular vesicles are separated without columns using an EasySep™ magnet. Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube. Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectometry.​
Catalog No. 50-197-4600 Supplier STEMCELL Technologies Supplier No. 17891
The EasySep™ Human Pan-Extracellular Vesicle Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection. Extracellular vesicles are labeled with Tetrameric Antibody Complexes recognizing CD9, CD81, and CD63, and magnetic particles. Labeled extracellular vesicles are separated without columns using an EasySep™ magnet. Unwanted biofluid components are simply poured off, while desired extracellular vesicles remain in the tube. Isolated extracellular vesicles are available for downstream applications such as DNA/RNA extraction, Western blot, or mass spectometry.​