Learn More
STEMCELL Technologies EasySep™ Human PE Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974545
Encompass
The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Human PE Positive Selection Kit (Catalog #18551). For highly purified particle-free cells, we recommend the EasySep™ Release Human PE Positive Selection Kit (Catalog #17654) (https://www.stemcell.com/products/easysep-release-human-pe-positive-selection-kit.html)
Catalog No. 50-197-4545 Supplier STEMCELL Technologies Supplier No. 17664
The EasySep™ Human PE Positive Selection Kit II is designed to isolate cells that are labeled with PE (phycoerythrin)-conjugated antibodies by positive selection. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ Human PE Positive Selection Kit (Catalog #18551). For highly purified particle-free cells, we recommend the EasySep™ Release Human PE Positive Selection Kit (Catalog #17654) (https://www.stemcell.com/products/easysep-release-human-pe-positive-selection-kit.html)