Learn More
STEMCELL Technologies EasySep™ Human TCR Alpha/Beta Depletion Kit, undefined: 1 Kit
SDP
Catalog No. 501974565
Encompass
The EasySep™ Human TCR Alpha/Beta Depletion Kit is designed to deplete human T cell receptor alpha/beta+ (TCRab+) cells from leukapheresis samples or expanded TCRa/b knockout cell cultures. TCRab+ cells are targeted with antibody complexes and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off.
Catalog No. 50-197-4565 Supplier STEMCELL Technologies Supplier No. 17847
The EasySep™ Human TCR Alpha/Beta Depletion Kit is designed to deplete human T cell receptor alpha/beta+ (TCRab+) cells from leukapheresis samples or expanded TCRa/b knockout cell cultures. TCRab+ cells are targeted with antibody complexes and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off.