Learn More
STEMCELL Technologies EasySep™ Human Th17 Cell Enrichment Kit II, For processing: 2 x 10^9 cells
SDP
Catalog No. NC1431597
Encompass
The EasySep™ Human Th17 Enrichment Kit II is a two-step method designed to enrich for human Th17 cells from fresh peripheral blood mononuclear cells. First, CCR6+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the cells and unwanted non-CD4+ T cells, CD45RA+ cells, and CXCR3+ cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction is enriched for IL-17-producing Th17 cells.This product can be used in place of the EasySep™ Human Th17 Cell Enrichment Kit (Catalog #18162) for even faster cell isolations.
Catalog No. NC1431597 Supplier STEMCELL Technologies Supplier No. 17862
The EasySep™ Human Th17 Enrichment Kit II is a two-step method designed to enrich for human Th17 cells from fresh peripheral blood mononuclear cells. First, CCR6+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the cells and unwanted non-CD4+ T cells, CD45RA+ cells, and CXCR3+ cells are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction is enriched for IL-17-producing Th17 cells.This product can be used in place of the EasySep™ Human Th17 Cell Enrichment Kit (Catalog #18162) for even faster cell isolations.