Learn More
STEMCELL Technologies EasySep™ Magnet, For isolating: 1 Unit
SDP
Catalog No. NC9284020
Encompass
The EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents. The EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns. This magnet is designed to hold a standard 12 x 75 mm (5 mL) polystyrene tube.
Catalog No. NC9284020 Supplier STEMCELL Technologies Supplier No. 18000
The EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents. The EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns. This magnet is designed to hold a standard 12 x 75 mm (5 mL) polystyrene tube.