Learn More
STEMCELL Technologies Easy 50 EasySep™ Magnet, For isolating: 1 Unit
SDP
Catalog No. NC0322302
Encompass
The Easy 50 EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4 x 10^9 cells). The Easy 50 EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles without the use of columns. The Easy 50 EasySep™ Magnet is designed to hold a standard 50 mL conical tube.
Catalog No. NC0322302 Supplier STEMCELL Technologies Supplier No. 18002
The Easy 50 EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a large number of cells (up to 4 x 10^9 cells). The Easy 50 EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with magnetic EasySep™ particles without the use of columns. The Easy 50 EasySep™ Magnet is designed to hold a standard 50 mL conical tube.