Learn More
STEMCELL Technologies EasySep™ Mouse B Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0378722
Encompass
The EasySep™ Mouse B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-B cells and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using and EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For isolation of B cells expressing CD11b or CD43, we recommend using the EasySep™ Mouse Pan-B Cell Isolation kit (Catalog #19844 (https://www.stemcell.com/products/easysep-mouse-pan-b-cell-isolation-kit.html)).This product replaces the EasySep™ Mouse B Cell Enrichment Kit (Catalog #19754) for even faster cell isolations.
Catalog No. NC0378722 Supplier STEMCELL Technologies Supplier No. 19854
The EasySep™ Mouse B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-B cells and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using and EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For isolation of B cells expressing CD11b or CD43, we recommend using the EasySep™ Mouse Pan-B Cell Isolation kit (Catalog #19844 (https://www.stemcell.com/products/easysep-mouse-pan-b-cell-isolation-kit.html)).This product replaces the EasySep™ Mouse B Cell Enrichment Kit (Catalog #19754) for even faster cell isolations.