Learn More
STEMCELL Technologies EasySep™ Mouse CD117 (cKIT) Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. NC0059903
Encompass
The EasySep™ Mouse CD117 (cKIT) Positive Selection Kit is designed to isolate CD117+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labeled antibodies that is directed against CD117, which are then recognized by antibody complexes that are directed against PE and dextran. Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
Catalog No. NC0059903 Supplier STEMCELL Technologies Supplier No. 18757
The EasySep™ Mouse CD117 (cKIT) Positive Selection Kit is designed to isolate CD117+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labeled antibodies that is directed against CD117, which are then recognized by antibody complexes that are directed against PE and dextran. Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.