Learn More
STEMCELL Technologies EasySep™ Mouse CD11b Positive Selection Kit II, For processing: 2 x 10^9 cells
SDP
Catalog No. NC1028259
Encompass
The EasySep™ Mouse CD11b Positive Selection Kit II isolates highly purified CD11b+ cells from mouse splenocytes, bone marrow, lungs, brains or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.This product replaces the EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770) for even faster cell isolations and does not result in the labeling of isolated cells with PE.
Catalog No. NC1028259 Supplier STEMCELL Technologies Supplier No. 18970
The EasySep™ Mouse CD11b Positive Selection Kit II isolates highly purified CD11b+ cells from mouse splenocytes, bone marrow, lungs, brains or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.This product replaces the EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770) for even faster cell isolations and does not result in the labeling of isolated cells with PE.