Learn More
STEMCELL Technologies EasySep™ Mouse CD138 Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. NC1569842
Encompass
The EasySep™ Mouse CD138 Positive Selection Kit is designed to isolate CD138+ cells from mouse splenocytes, lymph nodes and bone marrow by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell culture, and hybridoma generation.
Catalog No. NC1569842 Supplier STEMCELL Technologies Supplier No. 18957
The EasySep™ Mouse CD138 Positive Selection Kit is designed to isolate CD138+ cells from mouse splenocytes, lymph nodes and bone marrow by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell culture, and hybridoma generation.