Learn More
STEMCELL Technologies EasySep™ Mouse CD90.1 Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. NC1704711
Encompass
The EasySep™ Mouse CD90.1 Positive Selection Kit is designed to isolate CD90.1+ (Thy1.1+) cells from single suspensions of splenocytes, lymph nodes and peripheral blood by positive selection. Desired cells are targeted with antibody complexes recognizing CD90.1 and dextran-coated magnetic particles. Labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
Catalog No. NC1704711 Supplier STEMCELL Technologies Supplier No. 18958
The EasySep™ Mouse CD90.1 Positive Selection Kit is designed to isolate CD90.1+ (Thy1.1+) cells from single suspensions of splenocytes, lymph nodes and peripheral blood by positive selection. Desired cells are targeted with antibody complexes recognizing CD90.1 and dextran-coated magnetic particles. Labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.