Learn More
STEMCELL Technologies EasySep™ Mouse Epithelial Cell Enrichment Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1657097
Encompass
The EasySep™ Mouse Epithelial Cell Enrichment Kit II is designed to isolate epithelial cells from freshly dissociated mouse mammary tissues by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted non-epithelial cells with biotinylated antibodies and magnetic particles. The magnetically labeled cells are separated from untouched desired cells by using an EasySep™ magnet and simply pouring the desired cells into a new tube.This product replaces the EasySep™ Epithelial Cell Enrichment Kit (Catalog #19758) for even faster isolations. It also replaces the cell enrichment portion of the EasySep™ Mouse Mammary Stem Cell Enrichment Kit (Catalog #19757). Additionally, Anti-Mouse CD24 Antibody, Clone M1/69, PE (Catalog #60099PE) and Anti-Mouse CD49f Antibody, Clone GoH3, FITC (Catalog #60037FI) are required to complete the kit.
Catalog No. NC1657097 Supplier STEMCELL Technologies Supplier No. 19868
The EasySep™ Mouse Epithelial Cell Enrichment Kit II is designed to isolate epithelial cells from freshly dissociated mouse mammary tissues by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted non-epithelial cells with biotinylated antibodies and magnetic particles. The magnetically labeled cells are separated from untouched desired cells by using an EasySep™ magnet and simply pouring the desired cells into a new tube.This product replaces the EasySep™ Epithelial Cell Enrichment Kit (Catalog #19758) for even faster isolations. It also replaces the cell enrichment portion of the EasySep™ Mouse Mammary Stem Cell Enrichment Kit (Catalog #19757). Additionally, Anti-Mouse CD24 Antibody, Clone M1/69, PE (Catalog #60099PE) and Anti-Mouse CD49f Antibody, Clone GoH3, FITC (Catalog #60037FI) are required to complete the kit.