Learn More
STEMCELL Technologies EasySep™ Mouse Pan-Naïve T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0648770
Encompass
The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells (CD11b, CD19, CD24, CD25, CD44, CD45R/B220, CD49b, TER119) and streptavidin-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.Click here (/easysep-15-minute-kits) to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.
Catalog No. NC0648770 Supplier STEMCELL Technologies Supplier No. 19848
The EasySep™ Mouse Pan-Naïve T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-naïve T cells (CD11b, CD19, CD24, CD25, CD44, CD45R/B220, CD49b, TER119) and streptavidin-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.Click here (/easysep-15-minute-kits) to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.