Learn More
STEMCELL Technologies EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0640704
Encompass
The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labeled with biotinylated antibodies. Labeled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet. Desired cells are untouched and poured off into a new tube.
Catalog No. NC0640704 Supplier STEMCELL Technologies Supplier No. 19860
The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labeled with biotinylated antibodies. Labeled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet. Desired cells are untouched and poured off into a new tube.