Learn More
STEMCELL Technologies EasySep™ Mouse Memory CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974717
Encompass
The EasySep™ Mouse Memory CD4+ T Cell Isolation Kit is designed to isolate pan-memory CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-memory CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD45R, CD45RB, CD49b, CD105 and TCRγ/δ) and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.Learn more about our next-generation EasySep™ mouse cell isolation kits (/easysep-15-minute-kits), featuring RapidSphere™ technology.
Catalog No. 50-197-4717 Supplier STEMCELL Technologies Supplier No. 19767
The EasySep™ Mouse Memory CD4+ T Cell Isolation Kit is designed to isolate pan-memory CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-memory CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD45R, CD45RB, CD49b, CD105 and TCRγ/δ) and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.Learn more about our next-generation EasySep™ mouse cell isolation kits (/easysep-15-minute-kits), featuring RapidSphere™ technology.