Learn More
STEMCELL Technologies EasySep™ Mouse Pan-B Cell Isolation Kit
SDP
Supplier:  STEMCELL Technologies 19844
Encompass
10-Minute cell isolation kit using immunomagnetic negative selection. The EasySep™ Mouse Pan-B Cell Isolation Kit is designed to isolate all B cells, including conventional B-2 B cells, B-1 B cells and plasma cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non pan-B cells (CD4, CD8, CD11c, CD49b, CD90.2, Ly-6C/G (Gr-1), TER119) and streptavidin-coated magnetic particles (RapidSpheres™ ). Labeled cells are separated using an EasySep™ magnet without the use of columns.This kit is compatible for use with cells from disease models where the malignant cells (B-CLL) express CD43 or CD11b. For isolation of conventional B cells only, we recommend using the EasySep™ Mouse B Cell Isolation Kit (Catalog #19854).Click here to learn about our next-generation EasySep™ mouse cell isolation kits, featuring RapidSphere™ technology.
Catalog No. NC0492710
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov