Learn More
STEMCELL Technologies EasySep™ Mouse SCA1 Positive Selection Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. 501974646
Encompass
The EasySep™ Mouse SCA1 Positive Selection Kit is designed to isolate SCA1+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labeled antibodies that is directed against SCA1, which are then recognized by antibody complexes that are directed against PE and dextran. Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This kit replaces the EasySep™ Mouse SCA1 Biotin Positive Selection Kit (Catalog #18856) for even faster cell isolations.
Catalog No. 50-197-4646 Supplier STEMCELL Technologies Supplier No. 18756
The EasySep™ Mouse SCA1 Positive Selection Kit is designed to isolate SCA1+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labeled antibodies that is directed against SCA1, which are then recognized by antibody complexes that are directed against PE and dextran. Labeled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This kit replaces the EasySep™ Mouse SCA1 Biotin Positive Selection Kit (Catalog #18856) for even faster cell isolations.