Learn More
STEMCELL Technologies EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1780403
Encompass
The EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit is designed to isolate CD11b+Gr1+ myeloid-derived suppressor cells (MDSCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-MDSCs and streptavidin-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell sorting, and cell culture.
Catalog No. NC1780403 Supplier STEMCELL Technologies Supplier No. 19867
The EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit is designed to isolate CD11b+Gr1+ myeloid-derived suppressor cells (MDSCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-MDSCs and streptavidin-coated magnetic particles (RapidSpheres™). Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell sorting, and cell culture.