Learn More
STEMCELL Technologies EasySep™ Multistand with EasySep™ Magnets, Size: 1 Unit
SDP
Catalog No. 501974625
Encompass
The EasySep™ Multistand allows the separation of up to 4 samples at one time. An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 "The Big Easy" EasySep™ Magnets (Catalog #18001). After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Catalog No. 50-197-4625 Supplier STEMCELL Technologies Supplier No. 18004
Add to Cart
Add to Cart
Request a Quote
The EasySep™ Multistand allows the separation of up to 4 samples at one time. An ingenious reversible clamping bar allows the Multistand to hold up to 4 EasySep™ (Catalog #18000) or 4 "The Big Easy" EasySep™ Magnets (Catalog #18001). After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.