Learn More
STEMCELL Technologies EasySep™ Human CD8+ T Cell Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0050243
Encompass
The EasySep™ Human CD8+ T Cell Enrichment Kit is designed to isolate CD8+ T cells from fresh or previously frozen peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis sample by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD8+ T cells and glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human CD8+ T Cell Isolation Kit (17953) (http://www.stemcell.com/en/Products/All-Products/EasySep-Human-CD8-T-Cell-Isolation-Kit.aspx) which isolates cells in just 8 minutes.
Catalog No. NC0050243 Supplier STEMCELL Technologies Supplier No. 19053
The EasySep™ Human CD8+ T Cell Enrichment Kit is designed to isolate CD8+ T cells from fresh or previously frozen peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis sample by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD8+ T cells and glycophorin A and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations, we recommend the new EasySep™ Human CD8+ T Cell Isolation Kit (17953) (http://www.stemcell.com/en/Products/All-Products/EasySep-Human-CD8-T-Cell-Isolation-Kit.aspx) which isolates cells in just 8 minutes.