Learn More
STEMCELL Technologies EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19582RF
Encompass
The EasySep™ Non-Human Primate CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This kit has been verified for use with rhesus and cynomolgus macaques.
Catalog No. 50-197-4694