Learn More
STEMCELL Technologies EasySep™ Non-Human Primate CD8+ T Cell Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974696
Encompass
The EasySep™ Non-Human Primate CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This kit has been verified for use with rhesus and cynomolgus macaques.
Catalog No. 50-197-4696 Supplier STEMCELL Technologies Supplier No. 19583RF
The EasySep™ Non-Human Primate CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from fresh peripheral blood mononuclear cells by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This kit has been verified for use with rhesus and cynomolgus macaques.