Learn More
STEMCELL Technologies EasySep™ Direct Human PBMC Isolation Kit, For processing: 100 mL whole blood processed
SDP
Catalog No. NC1636641
Encompass
The EasySep™ Direct Human PBMC Isolation Kit is designed to isolate highly purified peripheral blood mononuclear cells (PBMCs) from fresh whole blood, buffy coat, bone marrow, cord blood, or leukapheresis products by negative selection. Red blood cells (RBCs), platelets, and unwanted cells are targeted for removal with antibody complexes and magnetic particles and separated using an EasySep™ magnet. Untouched PBMCs are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction.
Catalog No. NC1636641 Supplier STEMCELL Technologies Supplier No. 19654
The EasySep™ Direct Human PBMC Isolation Kit is designed to isolate highly purified peripheral blood mononuclear cells (PBMCs) from fresh whole blood, buffy coat, bone marrow, cord blood, or leukapheresis products by negative selection. Red blood cells (RBCs), platelets, and unwanted cells are targeted for removal with antibody complexes and magnetic particles and separated using an EasySep™ magnet. Untouched PBMCs are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture, or DNA/RNA extraction.