Learn More
STEMCELL Technologies EasySep™ PE Positive Selection Kit II, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1718424
Encompass
The EasySep™ PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ PE Positive Selection Kit (Catalog #18557) for even faster cell isolations.
Catalog No. NC1718424 Supplier STEMCELL Technologies Supplier No. 17684
The EasySep™ PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ PE Positive Selection Kit (Catalog #18557) for even faster cell isolations.