Learn More
STEMCELL Technologies EasySep™ Rat IgM Depletion Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 501974639
Encompass
The EasySep™ Rat IgM Depletion Kit is designed to deplete IgM+ cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by positive selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Catalog No. 50-197-4639 Supplier STEMCELL Technologies Supplier No. 18644
The EasySep™ Rat IgM Depletion Kit is designed to deplete IgM+ cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by positive selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.