Learn More
STEMCELL Technologies EasySep™ RBC Depletion Reagent for RoboSep, For processing: 100 mL whole blood processed
SDP
Catalog No. 501974634
Encompass
The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection. RBCs are targeted with Tetrameric Antibody Complexes recognizing Glycophorin A and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube while desired cells are poured off.This product replaces the EasySep™ Human Glycophorin A Depletion Kit (Catalog #18352) for even faster cell isolations.
Catalog No. 50-197-4634 Supplier STEMCELL Technologies Supplier No. 18170RF
The EasySep™ RBC Depletion Reagent is designed to immunomagnetically deplete red blood cells (RBCs) from fresh human peripheral blood, umbilical cord blood, bone marrow or leukapheresis samples by negative selection. RBCs are targeted with Tetrameric Antibody Complexes recognizing Glycophorin A and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube while desired cells are poured off.This product replaces the EasySep™ Human Glycophorin A Depletion Kit (Catalog #18352) for even faster cell isolations.