Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer, Size: 100 mL
SDP
Catalog No. NC9037479
Encompass
EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood.
Catalog No. NC9037479 Supplier STEMCELL Technologies Supplier No. 20120
EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood.