Learn More
STEMCELL Technologies EasySep™ Release Human PE Positive Selection Kit, Size: 1 x 10^9 cells
SDP
Catalog No. NC1473310
Encompass
Immunomagnetic positive selection of particle-free human cells labeled with PE-conjugated antibodies
Catalog No. NC1473310 Supplier STEMCELL Technologies Supplier No. 17654
Immunomagnetic positive selection of particle-free human cells labeled with PE-conjugated antibodies