Learn More
STEMCELL Technologies EasySep™ Mouse Neutrophil Enrichment Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0268142
Encompass
The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils. Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran. These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. NC0268142 Supplier STEMCELL Technologies Supplier No. 19762
The EasySep™ Mouse Neutrophil Enrichment Kit is designed to isolate neutrophils from single-cell suspensions of bone marrow or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-neutrophils. Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran. These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.