Learn More
Interscience EasySpiral
SDP
Catalog No. 16949030 Shop All Interscience Products
Change view
Click to view available options
Product Line:
EasySpiral
EasySpiral Dilute
EasySpiral Pro
EasySpiral Pro Milk
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
16-949-030 EasySpiral
16-949-051 EasySpiral Pro
16-948-919 EasySpiral Pro Milk
16-948-888 EasySpiral Dilute
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16-949-030 Supplier Interscience Supplier No. 412000
Add to Cart
Add to Cart
Request a Quote
The easySpiral plater uses the Spiral technique for plating Petri dishes.
EasySpiral is an automatic plater of Petri dishes. It allows plating of a sample with 300 to 1.3x10^5 CFU/mL on one Petri dish, without prior dilution. Plated volumes are of 50 and 100 μL.
  • Syringe capacity: 1000 μL
  • Plating modes: exponential, circle
  • Counting range: from 300 to 1.3x10^5 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro is an automatic plater of Petri dishes. It allows plating of a sample of 100 to 1x10^7 CFU/mL on one Petri dish, without prior dilution. Traceability is ensured and volumes are programmable via USB connection.
  • Traceability and programmable volumes
  • Plating modes: exponential, constant, circle
  • Counting range: from 100 to 1x10^7 CFU/mL
  • Plating cycle time: 25 seconds
EasySpiral Pro Milk is an automatic Petri dish plater. It allows plating of a sample of 100 to 1x10^7 CFU/mL on a single Petri dish, without prior dilution. EasySpiral Pro Milk is particularly suitable for the dairy industry in analyzing raw materials and payment for milk.
  • Sample intake in the sample holder
  • Plating modes: exponential, pour, circle
  • Counting range: from 100 to 1x10^7 CFU mL
  • Plating cycle time: 25 seconds
EasySpiral Dilute is an automatic diluter and plater of Petri dishes. It allows 1/10th serial dilutions and plating up to 6 log. dilution on 1 single Petri dish. For a sample of 30 to 1 x 10^12 bacteria, no more manual dilutions and plating is automatic.
  • Automatic cycle of 5 successive dilutions at 1/10th
  • Plating modes: exponential, constant, circle, pour plate
  • Count range: from 30 to 1x10^12 CFU/mL
  • Plating cycle time: 134 seconds
Specifications
Type Automatic Plater
Capacity 1000 μL
Product Line EasySpiral
Speed 1 Cycle/25 sec.
Voltage 110/240 V